missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD279 (PD-1) Mouse anti-Human, PE-Cy7, Clone: eBioJ105 (J105), eBioscience™

Mouse Monoclonal Antibody

$209.88 - $460.68

Specifications

Antigen CD279 (PD-1)
Clone eBioJ105 (J105)
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123364 Life Technologies
LS25279942
100 Tests Each for $460.68
50245744 Life Technologies
LS25279941
25 Tests Each for $209.88
Specifications

Description

The J105 monoclonal antibody reacts with the human PD-1 (programmed death-1), a 55kDa member of the CD28 immunoglobulin superfamily. PD-1 contains the immunoreceptor tyrosine-based inhibitory motif (ITIM) and plays a key role in peripheral tolerance and autoimmune disease. PD-1 is expressed predominantly on activated T and B lymphocytes. Two novel members of the B7 family have been identified as the PD-1 ligands, PD-L1 (B7-H1) and PD-L2 (B7-DC). Evidence reported to date suggests overlapping functions for these two PD-1 ligands and their constitutive expression on some normal tissues and upregulation on activated antigen-presenting cells.
Costaining experiments suggest that eBioJ105 recognizes a different epitope than MIH4 (cat. 11-9969).
Description & Specifications

Specifications

CD279 (PD-1)
Murine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioJ105 (J105)
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.5μg)/test