Please login to your online account to display your discounted pricing

CD278 (ICOS) Rat anti-Mouse, PE-Cy5, Clone: 7E.17G9, eBioscience™

Rat Monoclonal Antibody

$216.05 - $382.80

Specifications

Antigen CD278 (ICOS)
Clone 7E.17G9
Host Species Rat
Gene Alias Inducible T-cell co-stimulator
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014156

 
life technologies
LS15994281
50μg Each for $216.05

5014157

 
life technologies
LS15994282
100μg Each for $382.80
Description & Specifications

Specifications

Antigen CD278 (ICOS)
Clone 7E.17G9
Host Species Rat
Gene Alias Inducible T-cell co-stimulator
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 7E.17G9 monoclonal antibody reacts with mouse ICOS (Inducible COStimulatory molecule), a T cell specific molecule and a third member of the CD28/CTLA-4 family. A homodimer of 47-57 kDa, ICOS is expressed on activated T cells, has potent costimulatory activity for T cell activation and proliferation and is required for humoral immune response. ICOS binds to its ligand on activated APC including B cells called B7h/B7RP-1 and is thought to play a protective role in inflammatory autoimmune diseases. ICOS may be involved in the development of Th2 cells.