missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD278 (ICOS) Rat anti-Mouse, Clone: 7E.17G9, eBioscience™

Rat Monoclonal Antibody

$104.28 - $341.88

Specifications

Antigen CD278 (ICOS)
Clone 7E.17G9
Host Species Rat
Gene Alias Inducible T-cell co-stimulator
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013901 Life Technologies
LS14994281
50μg Each for $104.28
5013902 Life Technologies
LS14994285
500μg Each for $341.88
Specifications

Description

The 7E.17G9 monoclonal antibody reacts with mouse ICOS (Inducible COStimulatory molecule), a T cell specific molecule and a third member of the CD28/CTLA-4 family. A homodimer of 47-57 kDa, ICOS is expressed on activated T cells, has potent costimulatory activity for T cell activation and proliferation and is required for humoral immune response. ICOS binds to its ligand on activated APC including B cells called B7h/B7RP-1 and is thought to play a protective role in inflammatory autoimmune diseases. ICOS may be involved in the development of Th2 cells.
Description & Specifications

Specifications

CD278 (ICOS)
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
7E.17G9
Inducible T-cell co-stimulator
Flow Cytometry
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL