Please login to your online account to display your discounted pricing

CD278 (ICOS) Golden Syrian Hamster anti-Mouse, PerCP-eFluor 710, Clone: 15F9, eBioscience™

Golden Syrian Hamster Monoclonal Antibody

$158.05 - $390.05

Specifications

Antigen CD278 (ICOS)
Clone 15F9
Host Species Golden Syrian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50245950

 
life technologies
LS46994080
25μg Each for $158.05

50245951

 
life technologies
LS46994082
100μg Each for $390.05
Description & Specifications

Specifications

Antigen CD278 (ICOS)
Clone 15F9
Host Species Golden Syrian Hamster
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 15F9 monoclonal antibody reacts with mouse ICOS (Inducible COStimulatory molecule), a T cell specific molecule and a third member of the CD28/CTLA-4 family. A homodimer of 47-57 kDa, ICOS is expressed on activated T cells, has potent costimulatory activity for T cell activation and proliferation and is required for humoral immune response. IOS binds to its ligand on activated APC including B cells called B7h/B7RP-1 and is thought to play a protective role in inflammatory autoimmune diseases. ICOS may be involved in the development of Th2 cells.

The 15F9 antibody is reported to have functional activity.