missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD276 (B7-H3) Mouse anti-Human, PerCP-eFluor 710, Clone: 7-517, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $404.55

Specifications

Antigen CD276 (B7-H3)
Clone 7-517
Host Species Murine
Gene Alias B7 homolog 3
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123797 Life Technologies
LS46276941
25 Tests Each for $158.05
501130797 Life Technologies
LS46276942
100 Tests Each for $404.55
Specifications

Description

This 7-517 monoclonal antibody reacts with human CD276, which is also known as B7-H3. This type I transmembrane protein is expressed on immature and mature dendritic cells but not monocytes, granulocytes, or resting lymphocytes. Moreover, CD276 is expressed on non-hematopoietic cells such as osteoblasts, fibroblasts, and epithelial cells. However, CD276 expression can be induced on dendritic cells, monocytes, T, B, and NK cells. This co-stimulatory molecule binds activated T cells, leading to cell proliferation and IFN-gamma production.
Description & Specifications

Specifications

CD276 (B7-H3)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
7-517
B7 homolog 3
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.125μg)/test