Please login to your online account to display your discounted pricing

CD275 (B7-H2) Rat anti-Mouse, eFluor 660, Clone: HK5.3, eBioscience™

Rat Monoclonal Antibody

$158.05 - $375.55

Specifications

Antigen CD275 (B7-H2)
Clone HK5.3
Host Species Rat
Gene Alias B7RP1, ICOSL, GL50
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124414

 
life technologies
LS50598580
25μg Each for $158.05

501124415

 
life technologies
LS50598582
100μg Each for $375.55
Description & Specifications

Specifications

Antigen CD275 (B7-H2)
Clone HK5.3
Host Species Rat
Gene Alias B7RP1, ICOSL, GL50
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The HK5.3 monoclonal antibody reacts with mouse B7RP-1, also known as B7h, B7-H2, GL50 and ICOS Ligand. B7RP-1, a member of the B7 family, has a predicted molecular weight of ∽40kDa and belongs to the Ig superfamily. Mouse B7RP-1 is expressed by antigen presenting cells including B cells, monocytes/macrophages and dendritic cells. B7RP-1 binds to the ICOS molecule (AILIM, CRP-1) expressed by activated T cells. The interaction of ICOS/B7RP-1 plays an important role in the T cell costimulation pathway.