Please login to your online account to display your discounted pricing

CD275 (B7-H2) Mouse anti-Human, Biotin, Clone: MIH12, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $303.05

Specifications

Antigen CD275 (B7-H2)
Clone MIH12
Host Species Murine
Gene Alias B7RP-1, ICOS-L, GL50
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011999

 
life technologies
LS13588980
25μg Each for $129.05

5012000

 
life technologies
LS13588982
100μg Each for $303.05
Description & Specifications

Specifications

Antigen CD275 (B7-H2)
Clone MIH12
Host Species Murine
Gene Alias B7RP-1, ICOS-L, GL50
Species Reactivity Human
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The MIH12 monoclonal antibody reacts with human B7RP-1, also known as B7h, B7-H2, GL50 and ICOS Ligand. B7RP-1, a member of the B7 family, has a predicted molecular weight of approximately 40kDa and belongs to the Ig superfamily. Human B7RP-1 is expressed by activated monocytes/macrophages. B7RP-1 binds to the ICOS molecule (AILIM, CRP-1) expressed by activated T cells. The interaction of ICOS/B7RP-1 plays an important role in the T cell costimulation pathway.