Please login to your online account to display your discounted pricing

CD275 (B7-H2) Mouse anti-Human, APC, Clone: MIH12, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $361.05

Specifications

Antigen CD275 (B7-H2)
Clone MIH12
Host Species Mouse
Gene Alias B7RP-1, ICOS-L, GL50
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123156

 
affymetrix
17588941
25 tests Each for $187.05

501129724

 
affymetrix
17588942
100 tests Each for $361.05
Description & Specifications

Specifications

Antigen CD275 (B7-H2)
Clone MIH12
Host Species Mouse
Gene Alias B7RP-1, ICOS-L, GL50
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate APC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The MIH12 monoclonal antibody reacts with human B7RP-1, also known as B7h, B7-H2, GL50 and ICOS Ligand. B7RP-1, a member of the B7 family, has a predicted molecular weight of approximately 40kDa and belongs to the Ig superfamily. Human B7RP-1 is expressed by activated monocytes/macrophages. B7RP-1 binds to the ICOS molecule (AILIM, CRP-1) expressed by activated T cells. The interaction of ICOS/B7RP-1 plays an important role in the T cell costimulation pathway.