missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD274 (PD-L1, B7-H1) Rat anti-Mouse, Super Bright 645, Clone: MIH5, eBioscience™ DFS Item

Rat Monoclonal Antibody

$233.00 - $496.00

Specifications

Antigen CD274 (PD-L1, B7-H1)
Clone MIH5
Target Species Mouse
Conjugate Super Bright 645
Isotype IgG2a, lambda
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
64598280 View Documents Life Technologies
LS64598280
25 μg Each for $233.00
 
64598282 View Documents Life Technologies
LS64598282
100 μg Each for $496.00
 
Description

Description

Description: The 11-26c monoclonal antibody reacts with the delta heavy chain of mouse IgD. It does not react with other classes of mouse immunoglobulin including IgA, IgG or IgM. IgD is expressed by peripheral mature B cells. 11-26c does not activate B cells. Applications Reported: This 11-26c (11-26) antibody has been reported for use in flow cytometric analysis. Applications Tested: This 11-26c (11-26) antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 645 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 645 nm. We recommend using a 660/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No. SB-4400) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation.

Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No. 00-5333) for up to 3 days in the dark at 4 degrees C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405 nm; Emission: 645 nm; Laser: Violet Laser Programmed death receptor ligand 1 (PD-L1, also called B7-H1) is a recently described B7 family member. To date, one specific receptor has been identified that can be ligated by PD-L1. This receptor, programmed death receptor 1 (PD-1), has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production. PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion.
Specifications

Specifications

CD274 (PD-L1, B7-H1)
Mouse
IgG2a, lambda
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
MIH5
Super Bright 645
Flow Cytometry
Primary
Monoclonal
PBS with BSA and 0.09% sodium azide; pH 7.2
RUO