Please login to your online account to display your discounted pricing

CD274 (PD-L1, B7-H1) Mouse anti-Human, PE, Clone: MIH1, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $377.00

Specifications

Antigen CD274 (PD-L1, B7-H1)
Clone MIH1
Host Species Mouse
Gene Alias PD-L1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011054

 
life technologies
LS12598341
25 tests Each for $187.05

5011055

 
life technologies
LS12598342
100 tests Each for $377.00
Description & Specifications

Specifications

Antigen CD274 (PD-L1, B7-H1)
Clone MIH1
Host Species Mouse
Gene Alias PD-L1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The MIH1 monoclonal antibody reacts with human B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH1 is reported to be a blocking antibody.