Please login to your online account to display your discounted pricing

CD274 (PD-L1, B7-H1) Mouse anti-Human, Clone: MIH1, eBioscience™

Mouse Monoclonal Antibody

$85.55 - $158.05

Specifications

Antigen CD274 (PD-L1, B7-H1)
Clone MIH1
Host Species Murine
Gene Alias PD-L1,
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122705

 
life technologies
LS14598380
25μg Each for $85.55

5013404

 
life technologies
LS14598382
100μg Each for $158.05
Description & Specifications

Specifications

Antigen CD274 (PD-L1, B7-H1)
Clone MIH1
Host Species Murine
Gene Alias PD-L1,
Species Reactivity Human
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The MIH1 monoclonal antibody reacts with human B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH1 is reported to be a blocking antibody.