missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD274 (B7-H1) Rat anti-Mouse, Clone: MIH5, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$143.88 - $434.28

Specifications

Antigen CD274 (B7-H1)
Clone MIH5
Host Species Rat
Gene Alias B7H1, PD-L1, PDL-1
Applications Flow Cytometry,Functional Assay
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014663 Life Technologies
LS16598281
50μg Each for $143.88
5014664 Life Technologies
LS16598282
100μg Each for $289.08
5014665 Life Technologies
LS16598285
500μg Each for $434.28
Specifications

Description

The MIH5 monoclonal antibody reacts with mouse B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes including T, B, NK and DC. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH5 is reported to be a blocking antibody.
Description & Specifications

Specifications

CD274 (B7-H1)
Rat
Flow Cytometry,Functional Assay
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL
MIH5
B7H1, PD-L1, PDL-1
RUO
Purified
Primary
aqueous buffer, no sodium azide