missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD274 (B7-H1) Rat anti-Mouse, Clone: 1-111A, eBioscience™

Rat Monoclonal Antibody

$158.05 - $274.05

Specifications

Antigen CD274 (B7-H1)
Clone 1-111A
Host Species Rat
Gene Alias B7H1, PD-L1, PDL-1, PDL1
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013941 Life Technologies
LS14997181
50μg Each for $158.05
5013942 Life Technologies
LS14997182
100μg Each for $274.05
Specifications

Description

The 1-111A monoclonal antibody reacts with mouse B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes including T, B, NK and DC. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. Preliminary testing suggests that 1-111A and MIH5 (Cat. No. 14-5982) cross-block each other.
Description & Specifications

Specifications

CD274 (B7-H1)
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
1-111A
B7H1, PD-L1, PDL-1, PDL1
Flow Cytometry
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL