missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD274 (B7-H1) Rat anti-Mouse, Biotin, Clone: MIH5, eBioscience™

Rat Monoclonal Antibody

$130.68 - $421.08

Specifications

Antigen CD274 (B7-H1)
Clone MIH5
Host Species Rat
Gene Alias B7H1, PD-L1, PDL-1
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122565 Life Technologies
LS13598285
500μg Each for $421.08
5012088 Life Technologies
LS13598281
50μg Each for $130.68
5012089 Life Technologies
LS13598282
100μg Each for $262.68
Specifications

Description

The MIH5 monoclonal antibody reacts with mouse B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes including T, B, NK and DC. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH5 is reported to be a blocking antibody.
Description & Specifications

Specifications

CD274 (B7-H1)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MIH5
B7H1, PD-L1, PDL-1
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL