missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD274 (B7-H1) Mouse anti-Human, APC, Clone: MIH1, eBioscience™

Mouse Monoclonal Antibody

$187.44 - $394.68

Specifications

Antigen CD274 (B7-H1)
Clone MIH1
Host Species Murine
Gene Alias PD-L1,
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015281 Life Technologies
LS17598341
25 Tests Each for $187.44
5015282 Life Technologies
LS17598342
100 Tests Each for $394.68
Specifications

Description

The MIH1 monoclonal antibody reacts with human B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH1 is reported to be a blocking antibody.
Description & Specifications

Specifications

CD274 (B7-H1)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MIH1
PD-L1,
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test