missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD273 (B7-DC) Mouse anti-Human, PerCP-eFluor 710, Clone: MIH18, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $434.28

Specifications

Antigen CD273 (B7-DC)
Clone MIH18
Host Species Murine
Gene Alias PD-L2
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123858 Life Technologies
LS46588841
25 Tests Each for $170.28
501128778 Life Technologies
LS46588842
100 Tests Each for $434.28
Specifications

Description

The MIH18 monoclonal antibody reacts with human CD273, also known as PD-L2 or B7-DC, a member of the B7 family that has a predicted molecular weight of approximately 25 kDa, and belongs to the Ig superfamily. B7-DC is primarily expressed by subpopulations of dendritic cells and monocytes/macrophages. Although B7-DC has structural and sequence similarities to the B7 family, it does not bind CD28/CTLA-4, rather it is a ligand for PD-1. The interactions between PD-1 and B7-DC/PD-L2 have been reported to be involved in costimulation or suppression of T cell proliferation depending on state of cellular activation. MIH18 is a useful tool for studies to elucidate the exact function of B7-DC/PD-L2 in APC/T cell interaction, and to characterize the expression pattern of this molecule in humans. MIH18 is a blocking antibody.
Description & Specifications

Specifications

CD273 (B7-DC)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
MIH18
PD-L2
Flow Cytometry
PerCP-eFluor 710
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.06μg)/test