Please login to your online account to display your discounted pricing

CD273 (B7-DC) Mouse anti-Human, PE, Clone: MIH18, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen CD273 (B7-DC)
Clone MIH18
Host Species Murine
Gene Alias PD-L2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010953

 
life technologies
LS12588841
25 Tests Each for $143.55

5010954

 
life technologies
LS12588842
100 Tests Each for $361.05
Description & Specifications

Specifications

Antigen CD273 (B7-DC)
Clone MIH18
Host Species Murine
Gene Alias PD-L2
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The MIH18 monoclonal antibody reacts with human B7-DC, also known as PD-L2. B7-DC, a recently identified member of the B7 family, has a predicted molecular weight of approximately 25kDa and belongs to the Ig superfamily. B7-DC is primarily expressed by subpopulations of dendritic cells and monocytes/macrophages. Although B7-DC has structural and sequence similarities to the B7 family, it does not bind CD28/CTLA-4, rather it is a ligand for PD-1. The interactions between PD-1 and B7-DC/PD-L2 have been reported to be involved in costimulation or suppression of T cell proliferation depending on state of cellular activation. MIH18 is a useful tool for studies to elucidate the exact function of B7-DC/PD-L2 in APC/T cell interaction and to characterize the expression pattern of this molecule in humans. MIH18 is a blocking antibody.