Please login to your online account to display your discounted pricing

CD272 (BTLA) Mouse anti-Mouse, APC, Clone: 8F4, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $448.05

Specifications

Antigen CD272 (BTLA)
Clone 8F4
Host Species Murine
Gene Alias B and T lymphocyte attenuator
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015265

 
life technologies
LS17595680
25μg Each for $143.55

5015266

 
life technologies
LS17595682
100μg Each for $448.05
Description & Specifications

Specifications

Antigen CD272 (BTLA)
Clone 8F4
Host Species Murine
Gene Alias B and T lymphocyte attenuator
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The BTLA antibody reacts with mouse BTLA, B and T lymphocyte attenuator from both BALB/c and C57Bl/6 strains. BTLA is expressed by peripheral lymphocytes, splenic macrophages, developing B cells in the bone marrow and developing T cells in thymus and mature, but not immature bone marrow-derived dendritic cells. BTLA is implicated as a negative regulator of the activation and/or function of various hemopoietic cell types. It is reported that BTLA binds to B7-H4, but further studies are needed to confirm this interaction.

Note: The anti-mouse BTLA monoclonal antibody 6F7 is reported to stain CD4+ and CD8+ single-positive (SP) thymocytes (Hurchla et al). However, other anti-mouse BTLA clones generated simultaneously with 6F7 (8F4, 3F9.D12, 6G3 and 6H6) do not stain SP thymocytes . It is not understood why there is a discrepancy in thymocyte staining. However, in staining of splenocytes and bone marrow cells, clones 8F4, 3F9.D12, 6G3 and 6H6 stain similar populations to 6F7.