Please login to your online account to display your discounted pricing

CD27 Mouse anti-Human, PE-Cy7, Clone: O323, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $442.25

Specifications

Antigen CD27
Clone O323
Host Species Mouse
Gene Alias TNFRSF7
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015489

 
life technologies
LS25027941
25 tests Each for $216.05

5015490

 
life technologies
LS25027942
100 tests Each for $442.25
Description & Specifications

Specifications

Antigen CD27
Clone O323
Host Species Mouse
Gene Alias TNFRSF7
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy7
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The O323 monoclonal antibody reacts with human CD27, a lymphocyte-specific member of the TNFR superfamily. CD27 is expressed by a subset of thymocytes and virtually all mature T cells and is upregulated upon T-cell stimulation. CD27 binds to CD70, and through this interaction, plays an important role in T cell-B cell interaction.