missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD27 Mouse anti-Human, APC, Clone: O323, eBioscience™

Mouse Monoclonal Antibody

$183.48 - $381.48

Specifications

Antigen CD27
Clone O323
Host Species Murine
Gene Alias TNFRSF7
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014929 Life Technologies
LS17027941
25 Tests Each for $183.48
5014930 Life Technologies
LS17027942
100 Tests Each for $381.48
Specifications

Description

The O323 monoclonal antibody reacts with human CD27, a lymphocyte-specific member of the TNFR superfamily. CD27 is expressed by a subset of thymocytes and virtually all mature T cells and is upregulated upon T-cell stimulation. CD27 binds to CD70, and through this interaction, plays an important role in T cell-B cell interaction.
Description & Specifications

Specifications

CD27
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
O323
TNFRSF7
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test