Please login to your online account to display your discounted pricing

CD27 Mouse anti-Human, Alexa Fluor 700, Clone: O323, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $435.00

Specifications

Antigen CD27
Clone O323
Host Species Murine
Gene Alias TNFRSF7
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124678

 
life technologies
LS56027941
25 Tests Each for $216.05

5016883

 
life technologies
LS56027942
100 Tests Each for $435.00
Description & Specifications

Specifications

Antigen CD27
Clone O323
Host Species Murine
Gene Alias TNFRSF7
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Alexa Fluor 700
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The O323 monoclonal antibody reacts with human CD27, a lymphocyte-specific member of the TNFR superfamily. CD27 is expressed by a subset of thymocytes and virtually all mature T cells and is upregulated upon T-cell stimulation. CD27 binds to CD70, and through this interaction, plays an important role in T cell-B cell interaction.