missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD27 Armenian Hamster, PE-Cy7, Clone: LG.7F9, eBioscience™

Armenian Hamster Monoclonal Antibody

$183.48 - $434.28

Specifications

Antigen CD27
Clone LG.7F9
Host Species Armenian Hamster
Gene Alias TNFRSF7
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123275 Life Technologies
LS25027182
100μg Each for $434.28
5015487 Life Technologies
LS25027180
25μg Each for $183.48
Specifications

Description

The LG.7F9 monoclonal antibody reacts with mouse CD27, a lymphocyte-specific member of the TNFR superfamily. CD27 is expressed by virtually all mature T cells and by a subpopulation of B cells, mainly memory B cells. In mouse, CD27 has been found on nearly all thymocytes excluding a population of CD46-CD8- precursors. CD27 binds to CD70 and, through this interaction, plays an important role in T cell-B cell interaction. It has been reported that triggering CD27 plays an important role in the maturation of CD4+ and CD8+ effector cells. LG.7F9 cross-reacts with human and rat CD27.
Description & Specifications

Specifications

CD27
Armenian Hamster
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL
LG.7F9
TNFRSF7
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer