Please login to your online account to display your discounted pricing

CD27 Armenian Hamster, PE, Clone: LG.7F9, eBioscience™

Armenian Hamster Monoclonal Antibody

$162.40 - $456.75

Specifications

Antigen CD27
Clone LG.7F9
Host Species Armenian Hamster
Gene Alias TNFRSF7
Species Reactivity Human, Mouse, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010297

 
life technologies
LS12027181
50μg Each for $162.40

5010298

 
life technologies
LS12027182
100μg Each for $263.90

5010299

 
life technologies
LS12027183
200μg Each for $456.75
Description & Specifications

Specifications

Antigen CD27
Clone LG.7F9
Host Species Armenian Hamster
Gene Alias TNFRSF7
Species Reactivity Human, Mouse, Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The LG.7F9 monoclonal antibody reacts with mouse CD27, a lymphocyte-specific member of the TNFR superfamily. CD27 is expressed by virtually all mature T cells and by a subpopulation of B cells, mainly memory B cells. In mouse, CD27 has been found on nearly all thymocytes excluding a population of CD46-CD8- precursors. CD27 binds to CD70 and, through this interaction, plays an important role in T cell-B cell interaction. It has been reported that triggering CD27 plays an important role in the maturation of CD4+ and CD8+ effector cells. LG.7F9 cross-reacts with human and rat CD27.