missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD268 (BAFF Receptor) Rat anti-Mouse, APC, Clone: eBio7H22-E16, eBioscience™

Rat Monoclonal Antibody

$170.28 - $368.28

Specifications

Antigen CD268 (BAFF Receptor)
Clone eBio7H22-E16
Host Species Rat
Gene Alias BAFFR, BAFF-R, TNFRSF13C, BCMD, BR3, BlySR3
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015261 Life Technologies
LS17594380
25μg Each for $170.28
5015262 Life Technologies
LS17594382
100μg Each for $368.28
Specifications

Description

The monoclonal antibody eBio7H22.E16 reacts with mouse BAFF-Receptor, a type III transmembrane TNF receptor family member. BAFF-R was found to be the predominant BAFF receptor expressed on peripheral B cells, in both humans and mice. The receptor is found on the surface of all B220+ B cells found in spleen. BAFF-R is expressed on activated/memory subsets of T-cells and is important for splenic B cell maturation and survival and is a major mediator of BAFF- dependent costimulatory responses in peripheral B and T cells. BAFF-R can cause a high level of autoimmune disease and a defect in BAFF-R will cause a decrease in generating mature B-Cells The ligand for the BAFF-R is BAFF also known as BLyS.
Description & Specifications

Specifications

CD268 (BAFF Receptor)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBio7H22-E16
BAFFR, BAFF-R, TNFRSF13C, BCMD, BR3, BlySR3
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL