missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD268 (BAFF Receptor) Mouse anti-Human, PE, Clone: 8A7, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $368.28

Specifications

Antigen CD268 (BAFF Receptor)
Clone 8A7
Host Species Murine
Gene Alias BR3, BLyS receptor 3, BAFF Receptor, TNFRSF13C
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011252 Life Technologies
LS12911741
25 Tests Each for $170.28
5011253 Life Technologies
LS12911742
100 Tests Each for $368.28
Specifications

Description

The 8A7 monoclonal antibody reacts with the human BAFF receptor, also known as B lymphocyte stimulator (BLyS protein) receptor, which is expressed on B cells. The ligand for this receptor, BAFF (B cell-activating factor of the TNF family) is a B cell survival factor and regulates CD21/35 and CD23 expression. Interaction of this ligand with its receptor causes elevated CD21/35 and CD23 expression, whereas receptor blockade has been shown to down-modulate expression.
Description & Specifications

Specifications

CD268 (BAFF Receptor)
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
8A7
BR3, BLyS receptor 3, BAFF Receptor, TNFRSF13C
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.00375μg)/test