missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD267 (TACI) Mouse anti-Human, PE, Clone: 11H3, eBioscience™

Mouse Monoclonal Antibody

$196.68 - $368.28

Specifications

Antigen CD267 (TACI)
Clone 11H3
Host Species Murine
Gene Alias TNFRSF13B
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011256 Life Technologies
LS12921741
25 Tests Each for $196.68
5011257 Life Technologies
LS12921742
100 Tests Each for $368.28
Specifications

Description

The 11H3 antibody reacts with human TACI, also known as tumor necrosis factor receptor 13B. TACI, like BCMA, binds BAFF and April. TACI and its ligands regulate humoral immune responses, activate NF-kB and c-jun N-terminal kinase, and are involved in B cell associated autoimmune diseases. TACI mRNA and protein are found predominantly in B-cells and correlate with BLyS binding in a panel of B-cell lines. TACI interacts with nanomolar affinity with the BLyS-related tumor necrosis factor homologue APRIL for which no clear in vivo role has been described. 11H3 does not block binding of BAFF/BLyS to TACI.
Description & Specifications

Specifications

CD267 (TACI)
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
11H3
TNFRSF13B
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer