Please login to your online account to display your discounted pricing

CD262 (DR5) Mouse anti-Human, Clone: DJR2-4 (a.k.a. 7-8), eBioscience™

Mouse Monoclonal Antibody

$143.55 - $288.55

Specifications

Antigen CD262 (DR5)
Clone DJR2-4 (a.k.a. 7-8)
Host Species Mouse
Gene Alias Trail-R2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013847

 
affymetrix
14990882
100μg Each for $288.55

5013846

 
affymetrix
14990880
25μg Each for $143.55
Description & Specifications

Specifications

Antigen CD262 (DR5)
Clone DJR2-4 (a.k.a. 7-8)
Host Species Mouse
Gene Alias Trail-R2
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The DJR2-4 monoclonal antibody reacts with human DR5, also known as TRAIL-R2, Apo2, TRICK2 and KILLER. DR5 binds to TRAIL, activates NF-κB, and induces TRAIL-mediated apoptosis. DR5 is expressed broadly by normal tissues as well as several tumor cells.