missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD262 (DR5) Mouse anti-Human, Clone: DJR2-2 (a.k.a. 2-6), eBioscience™

Mouse Monoclonal Antibody

$130.68 - $262.68

Specifications

Antigen CD262 (DR5)
Clone DJR2-2 (a.k.a. 2-6)
Host Species Murine
Gene Alias Trail-R2
Applications Flow Cytometry,Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013848 Life Technologies
LS14990980
25μg Each for $130.68
5013849 Life Technologies
LS14990982
100μg Each for $262.68
Specifications

Description

The DJR2-2 monoclonal antibody reacts with human DR5, also known as TRAIL-R2, Apo2, TRICK2, and KILLER. DR5 binds to TRAIL, activates NF-κB, and induces TRAIL-mediated apoptosis. DR5 is expressed broadly by normal tissues as well as several tumor cells.
Description & Specifications

Specifications

CD262 (DR5)
Murine
Flow Cytometry,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
DJR2-2 (a.k.a. 2-6)
Trail-R2
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer