Please login to your online account to display your discounted pricing

CD262 (DR5) Armenian Hamster anti-Mouse, Clone: MD5-1, Functional Grade, eBioscience™

Armenian Hamster Monoclonal Antibody, Functional Grade

$187.05 - $506.05

Specifications

Antigen CD262 (DR5)
Clone MD5-1
Host Species Armenian Hamster
Gene Alias TRAIL-R2, TRAILR2, TNFRSF10B
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014601

 
life technologies
LS16588381
50μg Each for $187.05

5014602

 
life technologies
LS16588382
100μg Each for $317.55

5014603

 
life technologies
LS16588385
500μg Each for $506.05
Description & Specifications

Specifications

Antigen CD262 (DR5)
Clone MD5-1
Host Species Armenian Hamster
Gene Alias TRAIL-R2, TRAILR2, TNFRSF10B
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The MD5-1 monoclonal antibody reacts with mouse DR5, also known as TRAIL-R2, Apo2, TRICK2, and KILLER. DR5 binds to TRAIL, activates NF-κB, and induces TRAIL-mediated apoptosis. DR5 mRNA is expressed broadly by normal tissue and human antigen is also expressed by some tumor cell lines.