Please login to your online account to display your discounted pricing

CD262 (DR5) Armenian Hamster anti-Mouse, Biotin, Clone: MD5-1, eBioscience™

Armenian Hamster Monoclonal Antibody

$172.55 - $361.05

Specifications

Antigen CD262 (DR5)
Clone MD5-1
Host Species Armenian Hamster
Gene Alias TRAIL-R2, TRAILR2, TNFRSF10B
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011983

 
life technologies
LS13588381
50μg Each for $172.55

5011984

 
life technologies
LS13588382
100μg Each for $361.05
Description & Specifications

Specifications

Antigen CD262 (DR5)
Clone MD5-1
Host Species Armenian Hamster
Gene Alias TRAIL-R2, TRAILR2, TNFRSF10B
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The MD5-1 monoclonal antibody reacts with mouse DR5, also known as TRAIL-R2, Apo2, TRICK2, and KILLER. DR5 binds to TRAIL, activates NF-κB, and induces TRAIL-mediated apoptosis. DR5 mRNA is expressed broadly by normal tissue and human antigen is also expressed by some tumor cell lines.