missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD261 (DR4) Mouse anti-Human, Clone: DJR1, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $262.68

Specifications

Antigen CD261 (DR4)
Clone DJR1
Host Species Murine
Gene Alias TRAIL Receptor 1, TRAIL-R1, TRAILR1
Applications Flow Cytometry,Immunohistochemistry (Frozen)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013505 Life Technologies
LS14664480
25μg Each for $130.68
5013506 Life Technologies
LS14664482
100μg Each for $262.68
Specifications

Description

The DJR1 monoclonal antibody reacts with human DR4, also known as TRAIL-R1. DR4 binds to TRAIL, activates NF-κB, and induces TRAIL-mediated apoptosis. DR4 is expressed by a variety of human tumor cells and at very low level on human leukocytes.

Expression has also been found inside the cell on some cell lines (Zhang et al). This has been confirmed with intracellular staining of Jurkat cell line. Please contact tech@ebioscience.com for more information.
Description & Specifications

Specifications

CD261 (DR4)
Murine
Flow Cytometry,Immunohistochemistry (Frozen)
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
DJR1
TRAIL Receptor 1, TRAIL-R1, TRAILR1
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer