Please login to your online account to display your discounted pricing

CD26 Mouse anti-Human, PE, Clone: 2A6, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $317.55

Specifications

Antigen CD26
Clone 2A6
Host Species Murine
Gene Alias Dipeptidyl Peptidase IV, DPPIV, DPP4,
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010296

 
life technologies
LS12026942
100 Tests Each for $317.55

501122221

 
life technologies
LS12026941
25 Tests Each for $143.55
Description & Specifications

Specifications

Antigen CD26
Clone 2A6
Host Species Murine
Gene Alias Dipeptidyl Peptidase IV, DPPIV, DPP4,
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The 2A6 monoclonal antibody reacts with human CD26, a 110kDa type II transmembrane dimer. CD26 is an ectoenzyme with dipeptidyl peptidase IV activity expressed by T cells and is upregulated upon T cell stimulation. It is thought that CD26 plays a role in interaction with the extracellular matrix molecules and in T cell activation.