Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD26/DPP IV/ADA-Binding Protein Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunofluorescence and flow cytometry experiments.

$457.53 - $1,014.96

Specifications

Antigen CD26/DPP IV/ADA-Binding Protein Ab-1
Clone 202-36
Applications Flow Cytometry
Applications Immunofluorescence
Applications Flow Cytometry
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS524P

 
thermo scientific™ lab vision™
MS524P
200μg/mL 1mL Each for $902.53

MS524P0

 
thermo scientific™ lab vision™
MS524P0
200μg/mL 100µL Each for $457.53

MS524P1

 
thermo scientific™ lab vision™
MS524P1
200μg/mL 500µL Each for $824.82

MS524P1ABX

 
thermo scientific™ lab vision™
MS524P1ABX
1mg/mL 100µL Each for $952.14

MS524PABX

 
thermo scientific™ lab vision™
MS524PABX
1mg/mL 200µL Each for $1,014.96
Description & Specifications

Specifications

Antigen CD26/DPP IV/ADA-Binding Protein Ab-1
Clone 202-36
Applications Flow Cytometry
Applications Immunofluorescence
Applications Flow Cytometry
Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Enzyme
Type Primary
Immunogen Human T cell clone
Isotype IgG2b kappa

Ensure accurate, reproducible results in immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD26/DPP IV/ADA-Binding Protein Ab-1, Mouse Monoclonal Antibody.

 

CD26 (also known as dipeptidyl peptidase IV [DPP IV], adenosine deaminase [ADA]-binding protein) is an atypical serine protease belonging to the prolyl oligopeptidase family. It is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. Cross-linking of CD26 using this antibody dramatically enhances the anti-CD3-induced IL-2 production.

Host Species: Mouse

Clone: 202-36

Isotype: IgG2b/κ

Species Reactivity: Human and Rat. Does not react with pig and sheep. Others not tested.

Epitope: Not determined

Immunogen: Human T-cell clone

Molecular Weight: 110 kDa

Positive Control: HEP-G2 cells, Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO