Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD26/DPP IV/ADA-Binding Protein Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunofluorescence and flow cytometry experiments.

Manufacturer: thermo scientific™ lab vision™  MS524P1ABX

 View more versions of this product

Catalog No. MS524P1ABX

  • / Each
 

Description & Specifications

Specifications

Immunogen Human T cell clone
Applications Flow Cytometry
Applications Immunofluorescence
Cross Reactivity Human
Cross Reactivity Rat
Quantity 100µL
Clone 202-36
Regulatory Status RUO
Format Concentrated
Isotype IgG2b kappa
Antigen CD26/DPP IV/ADA-Binding Protein Ab-1
Host Species Mouse
Conjugate Unlabeled
Research Discipline Enzyme
Classification Monoclonal
Type Primary
Concentration 1mg/mL

Ensure accurate, reproducible results in immunofluorescence and flow cytometry experiments with Thermo Scientific™ CD26/DPP IV/ADA-Binding Protein Ab-1, Mouse Monoclonal Antibody.

 

CD26 (also known as dipeptidyl peptidase IV [DPP IV], adenosine deaminase [ADA]-binding protein) is an atypical serine protease belonging to the prolyl oligopeptidase family. It is expressed on lymphocyte cells and is upregulated during T-cell activation. CD26 is also expressed on activated B cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. Cross-linking of CD26 using this antibody dramatically enhances the anti-CD3-induced IL-2 production.

Host Species: Mouse

Clone: 202-36

Isotype: IgG2b/κ

Species Reactivity: Human and Rat. Does not react with pig and sheep. Others not tested.

Epitope: Not determined

Immunogen: Human T-cell clone

Molecular Weight: 110 kDa

Positive Control: HEP-G2 cells, Lymph node or tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO