missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD258 (LIGHT) Mouse anti-Human, APC, Clone: 7-3 (7), eBioscience™

Mouse Monoclonal Antibody

$183.48 - $421.08

Specifications

Antigen CD258 (LIGHT)
Clone 7-3 (7)
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015133 Life Technologies
LS17258941
25 Tests Each for $183.48
5015134 Life Technologies
LS17258942
100 Tests Each for $421.08
Specifications

Description

This 7-3 (7) monoclonal antibody reacts with human CD258 (also known as LIGHT), a 29kDa type II transmembrane protein that is a member of the TNF ligand superfamily related to lymphotoxin-ß (LT-ß). Expressed on activated T cells, particularly CD8+ T cells, LIGHT plays a role in cell survival, inflammation, and tumor eradication. Furthermore, upon binding its ligand, herpesvirus entry mediator (HVEM), LIGHT induces a costimulatory effect on T cells, enhancing T cell proliferation and cytokine production. LIGHT has also been shown to indirectly interact with the LT-ß receptor.
Description & Specifications

Specifications

CD258 (LIGHT)
Murine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
7-3 (7)
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test