missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD254 (RANK Ligand) Rat anti-Mouse, Biotin, Clone: IK22/5, eBioscience™

Rat Monoclonal Antibody

$230.55 - $375.55

Specifications

Antigen CD254 (RANK Ligand)
Clone IK22/5
Host Species Rat
Gene Alias OPGL, TNFSF11, ODF
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012058 Life Technologies
LS13595281
50μg Each for $230.55
5012059 Life Technologies
LS13595282
100μg Each for $375.55
Specifications

Description

The IK22/5 monoclonal antibody reacts with mouse CD254 also known as TRANCE (TNF-related activation-induced cytokine receptor) and RANKL (receptor activator of NF-kappa B ligand) and OPGL (Osteoprotegerin Ligand). TRANCE is a type II membrane protein with predicted molecular weight of 35 kDa. Its extracellular domain is most closely related to TRAIL, FasL and TNF. TRANCE mRNA is upregulated by TCR stimulation and is expressed abundantly by mouse T cells not B cells in mouse thymus and lymph nodes. TRANCE-deficient mouse lacks osteoclasts. In addition, TRANCE has been shown to be involved in the interactions between T-dendritic and T-B cells. It was also found to be critical in osteoclast differentiation. The TRANCE-RANK interaction regulates the expression of the anti-apoptotic molecule Bcl-xL.
Description & Specifications

Specifications

CD254 (RANK Ligand)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
IK22/5
OPGL, TNFSF11, ODF
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL