Please login to your online account to display your discounted pricing

CD253 (TRAIL) Rat anti-Mouse, PE, Clone: N2B2, eBioscience™

Rat Monoclonal Antibody

$172.55 - $361.05

Specifications

Antigen CD253 (TRAIL)
Clone N2B2
Host Species Rat
Gene Alias APO2L, TNFSF10
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011010

 
life technologies
LS12595181
50μg Each for $172.55

5011011

 
life technologies
LS12595182
100μg Each for $361.05
Description & Specifications

Specifications

Antigen CD253 (TRAIL)
Clone N2B2
Host Species Rat
Gene Alias APO2L, TNFSF10
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The N2B2 monoclonal antibody reacts with mouse TNF-related apoptosis-inducing ligand (TRAIL), a member of the TNF superfamily. TRAIL is not detected on the surface of freshly isolated T, B, or NK cells, but can be induced preferentially on CD3- NK1.1+ NK cells after stimulation with IL-2 or IL-15. N2B2 inhibits IL-2- or IL-15-activated NK cell cytotoxicity against mouse fibrosarcoma L929 target cells.