missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD253 (TRAIL) Rat anti-Mouse, Clone: N2B2, eBioscience™

Rat Monoclonal Antibody

$143.88 - $394.68

Specifications

Antigen CD253 (TRAIL)
Clone N2B2
Host Species Rat
Gene Alias APO2L, TNFSF10
Applications Flow Cytometry,Functional Assay,Immunoprecipitation
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013350 Life Technologies
LS14595181
50μg Each for $143.88
5013351 Life Technologies
LS14595182
100μg Each for $262.68
5013352 Life Technologies
LS14595185
500μg Each for $394.68
Specifications

Description

The N2B2 monoclonal antibody reacts with mouse TNF-related apoptosis-inducing ligand (TRAIL), a member of the TNF superfamily. TRAIL is not detected on the surface of freshly isolated T, B, or NK cells, but can be induced preferentially on CD3- NK1.1+ NK cells after stimulation with IL-2 or IL-15. N2B2 inhibits IL-2- or IL-15-activated NK cell cytotoxicity against mouse fibrosarcoma L929 target cells.
Description & Specifications

Specifications

CD253 (TRAIL)
Rat
Flow Cytometry,Functional Assay,Immunoprecipitation
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
N2B2
APO2L, TNFSF10
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer