Please login to your online account to display your discounted pricing

CD252 (OX40 Ligand) Rat anti-Mouse, PerCP-eFluor 710, Clone: RM134L, eBioscience™

Rat Monoclonal Antibody

$129.05 - $332.05

Specifications

Antigen CD252 (OX40 Ligand)
Clone RM134L
Host Species Rat
Gene Alias Tumor necrosis factor ligand superfamily member 4, CD134 Ligand, gp34
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123870

 
life technologies
LS46590580
25μg Each for $129.05

501123871

 
life technologies
LS46590582
100μg Each for $332.05
Description & Specifications

Specifications

Antigen CD252 (OX40 Ligand)
Clone RM134L
Host Species Rat
Gene Alias Tumor necrosis factor ligand superfamily member 4, CD134 Ligand, gp34
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The RM134L monoclonal antibody reacts with mouse CD252 also known as OX-40 Ligand, a member of the TNF superfamily. OX-40L is induced on mouse splenic B cells stimulated with a combination of anti-IgM and anti-CD40. Neither resting nor activated mouse T cells express OX-40L. The interaction of OX-40 (CD134) with OX-40L has been implicated in T cell-dependent humoral response. RM134L inhibits the costimulatory activity of mouse OX-40L transfected cells for anti-CD3-stimulated T cell proliferation.