missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD25 Rat anti-Mouse, PE-Cy5.5, Clone: PC61.5, eBioscience™

Rat Monoclonal Antibody

$157.08 - $394.68

Specifications

Antigen CD25
Clone PC61.5
Host Species Rat
Gene Alias Interleukin-2 Receptor alpha
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123545 Life Technologies
LS35025180
25μg Each for $157.08
501129176 Life Technologies
LS35025182
100μg Each for $394.68
Specifications

Description

The PC61.5 antibody reacts with mouse CD25, the 55kDa interleukin-2 receptor alpha chain (IL-2R alpha). CD25 is expressed by early progenitors of the T and B lineage as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) to form the high affinity IL-2 receptor. Binding of IL-2 to both the high and low affinity classes of IL-2 receptor is inhibited by the PC61.5 antibody. CD25 plays a role in lymphocyte differentiation and activation/proliferation.
Description & Specifications

Specifications

CD25
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
PC61.5
Interleukin-2 Receptor alpha
Flow Cytometry
PE-Cy5.5
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL