missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD25 Rat anti-Mouse, PE, Clone: PC61.5, eBioscience™

Rat Monoclonal Antibody

$130.68 - $368.28

Specifications

Antigen CD25
Clone PC61.5
Host Species Rat
Gene Alias Interleukin-2 Receptor alpha
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122220 Life Technologies
LS12025181
50μg Each for $130.68
501129394 Life Technologies
LS12025183
200μg Each for $368.28
501129454 Life Technologies
LS12025182
100μg Each for $262.68
Specifications

Description

The PC61.5 antibody reacts with mouse CD25, the 55kDa interleukin-2 receptor alpha chain (IL-2R alpha). CD25 is expressed by early progenitors of the T and B lineage as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) to form the high affinity IL-2 receptor. Binding of IL-2 to both the high and low affinity classes of IL-2 receptor is inhibited by the PC61.5 antibody. CD25 plays a role in lymphocyte differentiation and activation/proliferation.
Description & Specifications

Specifications

CD25
Rat
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
PC61.5
Interleukin-2 Receptor alpha
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer