missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD25 Rat anti-Mouse, Biotin, Clone: eBio7D4 (7D4), eBioscience™

Rat Monoclonal Antibody

$85.55 - $578.55

Specifications

Antigen CD25
Clone eBio7D4 (7D4)
Host Species Rat
Gene Alias Interleukin-2 Receptor alpha
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011487 Life Technologies
LS13025281
50μg Each for $85.55
5011488 Life Technologies
LS13025282
100μg Each for $172.55
5011489 Life Technologies
LS13025285
500μg Each for $346.55
5011490 Life Technologies
LS13025286
1mg Each for $578.55
Specifications

Description

The eBio7D4 antibody reacts with mouse CD25, the 55kDa interleukin-2 receptor a chain (IL-2Ra). CD25 is expressed by early progenitors of the T and B lineage, as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) to form the high affinity IL-2 receptor. CD25 plays a role in lymphocyte differentiation and activation/proliferation. The epitopes recognized by eBio3C7, PC61.5 and eBio7D4 are all different allowing for depletion with one antibody (typically PC61.5) followed by subsequent detection with another one. eBio7D4 has been reported to block IL-2 binding but can be overcome by additional IL-2. The monoclonal antibody PC61.5 (cat. 16-0251) is recommended for in vivo studies.
Description & Specifications

Specifications

CD25
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBio7D4 (7D4)
Interleukin-2 Receptor alpha
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL