missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD25 Rat anti-Mouse, Alexa Fluor 700, Clone: PC61.5, eBioscience™

Rat Monoclonal Antibody

$118.80 - $266.64

Specifications

Antigen CD25
Clone PC61.5
Host Species Rat
Gene Alias Interleukin-2 Receptor alpha
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016880 Life Technologies
LS56025180
25μg Each for $118.80
5016881 Life Technologies
LS56025182
100μg Each for $266.64
Specifications

Description

The PC61.5 antibody reacts with mouse CD25, the 55kDa interleukin-2 receptor alpha chain (IL-2R alpha). CD25 is expressed by early progenitors of the T and B lineage as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) to form the high affinity IL-2 receptor. Binding of IL-2 to both the high and low affinity classes of IL-2 receptor is inhibited by the PC61.5 antibody. CD25 plays a role in lymphocyte differentiation and activation/proliferation.
Description & Specifications

Specifications

CD25
Rat
Flow Cytometry
Alexa Fluor 700
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
PC61.5
Interleukin-2 Receptor alpha
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer