Please login to your online account to display your discounted pricing

CD25 Mouse anti-Rat, Biotin, Clone: OX39, eBioscience™

Mouse Monoclonal Antibody

$114.55 - $346.55

Specifications

Antigen CD25
Clone OX39
Host Species Mouse
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra
Species Reactivity Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011526

 
affymetrix
13039081
50μg Each for $114.55

5011527

 
affymetrix
13039085
500μg Each for $346.55
Description & Specifications

Specifications

Antigen CD25
Clone OX39
Host Species Mouse
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra
Species Reactivity Rat
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The OX39 monoclonal antibody reacts with mouse CD25, the 55kDa interleukin-2 receptor alpha (IL-2R alpha). CD25 is expressed by early progenitors of the T and B lineages as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) to form the high affinity IL-2 receptor. CD25 plays a role in lymphocyte differentiation and activation/proliferation.

The OX39 clone has been shown to deplete CD25 cells in vivo.