missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD25 Mouse anti-Human, PE-Cy7, Clone: BC96, eBioscience™

Mouse Monoclonal Antibody

$196.68 - $394.68

Specifications

Antigen CD25
Clone BC96
Host Species Murine
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra, IL2Ra
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015485 Life Technologies
LS25025941
25 Tests Each for $196.68
5015486 Life Technologies
LS25025942
100 Tests Each for $394.68
Specifications

Description

The BC96 monoclonal antibody reacts with human CD25, the 55kDa interleukin-2 receptor α chain (IL-2Rα). CD25 is expressed by early progenitors of T and B lineage as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor β chain) and CD132 (common γ chain) to form the high affinity IL-2 receptor. CD25 plays a role in lymphocyte differentiating and activation/proliferation.
Description & Specifications

Specifications

CD25
Murine
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
5μL (0.25μg)/test
BC96
Interleukin-2 Receptor alpha, IL-2Ra, IL2Ra
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer