Please login to your online account to display your discounted pricing

CD25 Mouse anti-Human, PE-Cy5, Clone: BC96, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $433.55

Specifications

Antigen CD25
Clone BC96
Host Species Murine
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra, IL2Ra
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014033

 
life technologies
LS15025941
25 Tests Each for $216.05

5014034

 
life technologies
LS15025942
100 Tests Each for $433.55
Description & Specifications

Specifications

Antigen CD25
Clone BC96
Host Species Murine
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra, IL2Ra
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

The BC96 monoclonal antibody reacts with human CD25, the 55kDa interleukin-2 receptor alpha chain (IL-2Ralpha). CD25 is expressed by early progenitors of T and B lineage as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) to form the high affinity IL-2 receptor. CD25 plays a role in lymphocyte differentiating and activation/proliferation.