Please login to your online account to display your discounted pricing

CD25 Mouse anti-Human, PE, Clone: BC96, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $332.05

Specifications

Antigen CD25
Clone BC96
Host Species Murine
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra, IL2Ra
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010292

 
life technologies
LS12025941
25 Tests Each for $143.55

5010294

 
life technologies
LS12025980
25μg Each for $216.05

501129575

 
life technologies
LS12025942
100 tests Each for $332.05
Description & Specifications

Specifications

Antigen CD25
Clone BC96
Host Species Murine
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra, IL2Ra
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL; 5μL (0.125μg)/test

The BC96 monoclonal antibody reacts with human CD25, the 55kDa interleukin-2 receptor α chain (IL-2Rα). CD25 is expressed by early progenitors of T and B lineage as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor β chain) and CD132 (common γ chain) to form the high affinity IL-2 receptor. CD25 plays a role in lymphocyte differentiating and activation/proliferation.