Please login to your online account to display your discounted pricing

CD25 Mouse anti-Human, Non-human primate, PerCP-eFluor 710, Clone: CD25-4E3, eBioscience™

Mouse Monoclonal Antibody

$201.55 - $520.55

Specifications

Antigen CD25
Clone CD25-4E3
Host Species Mouse
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra
Species Reactivity Human, Non-human primate
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015939

 
life technologies
LS46025741
25 tests Each for $201.55

5015940

 
life technologies
LS46025742
100 tests Each for $520.55
Description & Specifications

Specifications

Antigen CD25
Clone CD25-4E3
Host Species Mouse
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra
Species Reactivity Human, Non-human primate
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.06μg)/test

This CD25-4E3 monoclonal antibody reacts to human CD25, which is also known as the low-affinity interleukin (IL)-2 receptor α. CD25 is expressed on a subet of T cells (many of which are regulatory T cells), activated T and B cells, and macrophages. This receptor subunit associates with CD122, the IL-2 receptor β chain, and CD132, the common γ chain, to form the high-affinity IL-2 receptor. IL-2 receptor signaling has been linked to cell proliferation and survival in lymphocytes. CD25 is a marker for Hodgkin’s disease, and has been associated with such autoimmune diseases as Type I diabetes.

This monoclonal antibody is cross-reactive to rhesus monkey.