Please login to your online account to display your discounted pricing

CD25 Mouse anti-Human, Non-human primate, PE-eFluor 610, Clone: CD25-4E3, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen CD25
Clone CD25-4E3
Host Species Mouse
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra
Species Reactivity Human, Non-human primate
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124747

 
life technologies
LS61025741
25 tests Each for $172.55

501124748

 
life technologies
LS61025742
100 tests Each for $433.55
Description & Specifications

Specifications

Antigen CD25
Clone CD25-4E3
Host Species Mouse
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra
Species Reactivity Human, Non-human primate
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-eFluor 610
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

This CD25-4E3 monoclonal antibody reacts to human CD25, which is also known as the low-affinity interleukin (IL)-2 receptor alpha. CD25 is expressed on activated T cells (including a subset of regulatory T cells), B cells, and macrophages. This receptor subunit associates with CD122, the IL-2 receptor beta chain, and CD132, the common gamma chain, to form the high-affinity IL-2 receptor. IL-2 receptor signaling has been linked to cell proliferation and survival in lymphocytes. CD25 is a marker for Hodgkin’s disease, and has been associated with such autoimmune diseases as Type I diabetes.

This monoclonal antibody is cross-reactive to rhesus monkey.