Please login to your online account to display your discounted pricing

CD25 Mouse anti-Human, Non-human primate, FITC, Clone: CD25-4E3, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $332.05

Specifications

Antigen CD25
Clone CD25-4E3
Host Species Murine
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra
Species Reactivity Human, Non-human primate
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122053

 
life technologies
LS11025741
25 Tests Each for $143.55

501122054

 
life technologies
LS11025742
100 Tests Each for $332.05
Description & Specifications

Specifications

Antigen CD25
Clone CD25-4E3
Host Species Murine
Gene Alias Interleukin-2 Receptor alpha, IL-2Ra
Species Reactivity Human, Non-human primate
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

This CD25-4E3 monoclonal antibody reacts to human CD25, which is also known as the low-affinity interleukin (IL)-2 receptor alpha. CD25 is expressed on activated T cells (including a subset of regulatory T cells), B cells, and macrophages. This receptor subunit associates with CD122, the IL-2 receptor beta chain, and CD132, the common gamma chain, to form the high-affinity IL-2 receptor. IL-2 receptor signaling has been linked to cell proliferation and survival in lymphocytes. CD25 is a marker for Hodgkin’s disease, and has been associated with such autoimmune diseases as Type I diabetes.

This monoclonal antibody is cross-reactive to rhesus monkey.